Assay Details
Target Gene Details
Entrez Gene ID: | 101929025 |
Gene Name: | long intergenic non-protein coding RNA 1516 |
Gene Aliases: |
- |
Location: |
Chr.10:25113058-25161233 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LINC01516 | NR_120649.1 | |||
DA753967.1 |