Assay Details
Target Gene Details
Entrez Gene ID: | 2074 |
Gene Name: | ERCC excision repair 6, chromatin remodeling factor |
Gene Aliases: |
ARMD5, CKN2, COFS, COFS1, CSB, POF11, RAD26, UVSS1 |
Location: |
Chr.10:49454480-49539538 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ERCC6 | NM_000124.3 | NP_000115.1 | ||
AB209504.1 | BAD92741.1 | |||
AK130100.1 | ||||
AK303022.1 | ||||
BC127104.1 | ||||
L04791.1 | AAA52397.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1051636 | Chr.10:49467246 - 49524234 on Build GRCh38 | Loss |
![]() ![]() |
nsv831867 | Chr.10:49399811 - 49580705 on Build GRCh38 | Gain |
![]() ![]() |
nsv1042677 | Chr.10:47993784 - 49928329 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)