Assay Details
Target Gene Details
Entrez Gene ID: | 105376488 |
Gene Name: | uncharacterized LOC105376488 |
Gene Aliases: |
- |
Location: |
Chr.10:33334437-33385325 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105376488 | XM_017017010.1 | 2 | 989 | XP_016872499.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831829 | Chr.10:33178304 - 33344113 on Build GRCh38 | Loss | LOC105376488 NRP1 |