Assay Details
Target Gene Details
Entrez Gene ID: | 54764 |
Gene Name: | zinc finger RANBP2-type containing 1 |
Gene Aliases: |
TRABID |
Location: |
Chr.10:124916896-124988059 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZRANB1 | XM_005269926.4 | XP_005269983.1 | ||
XM_017016357.1 | XP_016871846.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv552289 | Chr.10:124875780 - 124998761 on Build GRCh38 | Loss | CTBP2 ZRANB1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |