Assay Details
Target Gene Details
Entrez Gene ID: | 51684 |
Gene Name: | SUFU negative regulator of hedgehog signaling |
Gene Aliases: |
PRO1280, SUFUH, SUFUXL |
Location: |
Chr.10:102502879-102633458 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SUFU | NM_001178133.1 | NP_001171604.1 | ||
NM_016169.3 | NP_057253.2 | |||
XM_011539858.2 | XP_011538160.1 | |||
XM_011539860.2 | XP_011538162.1 | |||
XM_011539861.2 | XP_011538163.1 | |||
XM_011539863.2 | XP_011538165.1 | |||
XM_011539864.2 | XP_011538166.1 | |||
XM_017016323.1 | XP_016871812.1 | |||
AF144231.1 | AAF23890.1 | |||
AF159447.1 | AAF23893.1 | |||
AF172319.1 | AAD51655.1 | |||
AF175770.1 | AAD50501.1 | |||
AF222345.1 | AAF35866.1 | |||
AK312319.1 | ||||
AY358550.1 | AAQ88914.1 | |||
BC013291.2 | AAH13291.1 | |||
DA248208.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831968 | Chr.10:102482950 - 102664645 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv467445 | Chr.10:102501602 - 102627177 on Build GRCh38 | Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)