Assay Details
Target Gene Details
Entrez Gene ID: | 23590 |
Gene Name: | prenyl (decaprenyl) diphosphate synthase, subunit 1 |
Gene Aliases: |
COQ1, COQ10D2, DPS, SPS, TPRT, TPT, TPT 1, hDPS1 |
Location: |
Chr.10:26697052-26746798 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDSS1 | NM_001321978.1 | NP_001308907.1 | ||
NM_001321979.1 | NP_001308908.1 | |||
NM_014317.4 | NP_055132.2 | |||
XM_011519437.2 | XP_011517739.1 | |||
XM_017016011.1 | XP_016871500.1 | |||
AB209763.1 | BAD93000.1 | |||
AB210838.1 | BAE48216.1 | |||
AF118395.1 | AAD28559.1 | |||
AK024802.1 | ||||
AK223414.1 | BAD97134.1 | |||
BC049211.1 | AAH49211.1 | |||
BC063635.1 | AAH63635.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv550233 | Chr.10:26679242 - 26722394 on Build GRCh38 | Loss | PDSS1 |
esv3415799 | Chr.10:26640952 - 26944612 on Build GRCh38 | Duplication | PDSS1 LINC00202-1 ANKRD26 LINC00202-2 ABI1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |