Assay Details
Target Gene Details
Entrez Gene ID: | 101928687 |
Gene Name: | long intergenic non-protein coding RNA 1468 |
Gene Aliases: |
lnc-MBL2-4 |
Location: |
Chr.10:52450877-52470533 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LINC01468 | NR_120641.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv130e214 | Chr.10:52442194 - 52482055 on Build GRCh38 | Gain | LINC01468 |