Assay Details
Target Gene Details
Entrez Gene ID: | 4601 |
Gene Name: | MAX interactor 1, dimerization protein |
Gene Aliases: |
MAD2, MXD2, MXI, bHLHc11 |
Location: |
Chr.10:110207605-110287365 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MXI1 | NM_001008541.1 | NP_001008541.1 | ||
NM_005962.4 | NP_005953.4 | |||
NM_130439.3 | NP_569157.2 | |||
AK303949.1 | ||||
AK307090.1 | ||||
AK309718.1 | ||||
AL538051.3 | ||||
BC012907.1 | AAH12907.1 | |||
BC035128.1 | ||||
BG700036.1 | ||||
BM927870.1 | ||||
BT007069.1 | AAP35732.1 | |||
BX417051.2 | ||||
CR536576.1 | CAG38813.1 | |||
D63940.1 | BAA09972.1 | |||
KU178207.1 | ||||
KU178208.1 | ||||
KU178209.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1037615 | Chr.10:110229567 - 110260725 on Build GRCh38 | Loss | MXI1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |