Assay Details
Target Gene Details
Entrez Gene ID: | 3778 |
Gene Name: | potassium calcium-activated channel subfamily M alpha 1 |
Gene Aliases: |
BKTM, KCa1.1, MaxiK, SAKCA, SLO, SLO-ALPHA, SLO1, bA205K10.1, hSlo, mSLO1 |
Location: |
Chr.10:76869601-77637819 on Build GRCh38 |
Assay Gene Location: | Within Intron 37 |
Target Gene Details
Entrez Gene ID: | 101929328 |
Gene Name: | KCNMA1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:76888044-76978593 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KCNMA1-AS1 | NR_120655.1 | |||
BX092192.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |