Assay Details
Target Gene Details
Entrez Gene ID: | 57118 |
Gene Name: | calcium/calmodulin dependent protein kinase ID |
Gene Aliases: |
CKLiK, CaM-K1, CaMKID |
Location: |
Chr.10:12349514-12835545 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CAMK1D | NM_020397.3 | NP_065130.1 | ||
NM_153498.3 | NP_705718.1 | |||
XM_006717482.3 | XP_006717545.1 | |||
XM_006717483.3 | XP_006717546.1 | |||
XM_017016438.1 | XP_016871927.1 | |||
AB081726.1 | BAC19846.1 | |||
AF286366.1 | AAG00534.1 | |||
AK289393.1 | ||||
BC035745.1 | AAH35745.1 | |||
BU542765.1 | ||||
HY168807.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3622363 | Chr.10:12200775 - 12828318 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
esv2676569 | Chr.10:12231868 - 12393054 on Build GRCh38 | Deletion |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)