Assay Details
Target Gene Details
Entrez Gene ID: | 51008 |
Gene Name: | activating signal cointegrator 1 complex subunit 1 |
Gene Aliases: |
ASC1p50, CGI-18, SMABF2, p50 |
Location: |
Chr.10:72096032-72217134 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1161975 | Chr.10:72102274 - 72106890 on Build GRCh38 | Duplication | ASCC1 |
esv3623792 | Chr.10:72102264 - 72106318 on Build GRCh38 | Loss | ASCC1 |
nsv442192 | Chr.10:72102390 - 72106143 on Build GRCh38 | Loss | ASCC1 |
esv3891857 | Chr.10:72086126 - 72148450 on Build GRCh38 | Loss | SPOCK2 ASCC1 |
esv24033 | Chr.10:72102314 - 72106290 on Build GRCh38 | Loss | ASCC1 |
dgv154e199 | Chr.10:72102096 - 72106342 on Build GRCh38 | Deletion | ASCC1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |