Assay Details
Target Gene Details
Entrez Gene ID: | 5464 |
Gene Name: | pyrophosphatase (inorganic) 1 |
Gene Aliases: |
HEL-S-66p, IOPPP, PP, PP1, SID6-8061 |
Location: |
Chr.10:70202830-70233434 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPA1 | NM_021129.3 | NP_066952.1 | ||
AB026723.1 | BAA84702.1 | |||
AF092439.1 | AAP97214.1 | |||
AF108211.1 | AAD24964.1 | |||
AF119665.1 | AAF17222.1 | |||
AF154065.1 | AAD34643.1 | |||
AF217186.1 | AAG36780.1 | |||
AK058193.1 | ||||
BC001022.2 | ||||
BC061581.1 | ||||
BC105034.1 | AAI05035.1 | |||
BC105036.1 | AAI05037.1 | |||
BC107882.1 | AAI07883.1 | |||
BU520836.1 | ||||
EU794626.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv32737 | Chr.10:69504230 - 70786847 on Build GRCh38 | Gain+Loss | NPFFR1 SAR1A NEUROG3 TBATA TYSND1 ADAMTS14 LRRC20 C10orf35 TSPAN15 AIFM2 NODAL COL13A1 PALD1 H2AFY2 PRF1 PPA1 EIF4EBP2 |
nsv831908 | Chr.10:70210726 - 70389945 on Build GRCh38 | Gain | NPFFR1 LRRC20 PPA1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |