Assay Details
Target Gene Details
Entrez Gene ID: | 509 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial F1 complex, gamma polypeptide 1 |
Gene Aliases: |
ATP5C, ATP5CL1 |
Location: |
Chr.10:7788104-7807801 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ATP5C1 | NM_001001973.2 | NP_001001973.1 | ||
NM_001320886.1 | NP_001307815.1 | |||
NM_005174.3 | NP_005165.1 | |||
XM_017016290.1 | XP_016871779.1 | |||
BC000470.2 | AAH00470.1 | |||
BC000931.3 | AAH00931.1 | |||
BC013394.2 | AAH13394.1 | |||
BC016812.1 | AAH16812.1 | |||
BC020824.1 | AAH20824.1 | |||
BC026049.1 | AAH26049.1 | |||
D16562.1 | BAA03996.1 | |||
D16563.1 | BAA03997.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv549933 | Chr.10:7586456 - 8160993 on Build GRCh38 | Gain | GATA3-AS1 ITIH5 TAF3 GATA3 KIN ATP5C1 ITIH2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |