Assay Details
Target Gene Details
Entrez Gene ID: | 23268 |
Gene Name: | dynamin binding protein |
Gene Aliases: |
ARHGEF36, TUBA |
Location: |
Chr.10:99875571-100009953 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DNMBP | NM_015221.3 | NP_056036.1 | ||
XM_006717735.2 | XP_006717798.1 | |||
XM_011539559.2 | XP_011537861.1 | |||
AB023227.1 | BAA76854.1 | |||
AL833283.1 | ||||
AY196211.1 | AAP34307.1 |
Target Gene Details
Entrez Gene ID: | 100188954 |
Gene Name: | DNMBP antisense RNA 1 |
Gene Aliases: |
NCRNA00093, bA287G8.2 |
Location: |
Chr.10:99927209-99958998 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DNMBP-AS1 | NR_024130.2 | |||
AK098111.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831960 | Chr.10:99833844 - 99995448 on Build GRCh38 | Gain | DNMBP DNMBP-AS1 ABCC2 |
nsv552045 | Chr.10:99862766 - 100074335 on Build GRCh38 | Gain | CPN1 DNMBP DNMBP-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |