Assay Details
Target Gene Details
Entrez Gene ID: | 143279 |
Gene Name: | HECT domain E3 ubiquitin protein ligase 2 |
Gene Aliases: |
- |
Location: |
Chr.10:91409280-91514829 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HECTD2 | NM_001284274.1 | NP_001271203.1 | ||
NM_173497.3 | NP_775768.3 | |||
NM_182765.4 | NP_877497.3 | |||
NR_104291.1 | ||||
XM_017015750.1 | XP_016871239.1 | |||
XM_017015751.1 | XP_016871240.1 | |||
AK094625.1 | BAC04388.1 | |||
AK295723.1 | ||||
BC040187.1 | AAH40187.1 | |||
BX537856.1 | ||||
HY158536.1 |
Target Gene Details
Entrez Gene ID: | 100188947 |
Gene Name: | HECTD2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:91306962-91611460 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HECTD2-AS1 | NR_024467.1 | |||
DA898704.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1040403 | Chr.10:91406062 - 91465279 on Build GRCh38 | Gain |
![]() ![]() |
nsv7488 | Chr.10:91396533 - 91430108 on Build GRCh38 | Insertion |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)