Assay Details
Target Gene Details
Entrez Gene ID: | 27069 |
Gene Name: | growth hormone inducible transmembrane protein |
Gene Aliases: |
DERP2, HSPC282, MICS1, My021, PTD010, TMBIM5 |
Location: |
Chr.10:84139429-84153555 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GHITM | NM_014394.2 | NP_055209.2 | ||
AB009685.1 | BAA93049.1 | |||
AF060923.1 | AAG43135.1 | |||
AF078863.1 | AAD44495.1 | |||
AF131820.1 | AAD20052.1 | |||
AF161400.1 | AAF28960.1 | |||
AK098770.1 | ||||
AK130094.1 | ||||
AK130097.1 | ||||
AK225318.1 | ||||
AK292864.1 | ||||
AK297960.1 | ||||
AK298332.1 | ||||
AK299937.1 | ||||
AL136713.1 | CAB66648.1 | |||
AY358824.1 | AAQ89183.1 | |||
BC010354.1 | AAH10354.1 | |||
CR533519.1 | CAG38550.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1046749 | Chr.10:83270389 - 84629720 on Build GRCh38 | Gain | LRIT1 CDHR1 C10orf99 HOST2 LINC00858 LRIT2 GHITM LOC105378397 RGR CCSER2 |
nsv551745 | Chr.10:83260366 - 84632635 on Build GRCh38 | Gain | LRIT1 CDHR1 C10orf99 HOST2 LINC00858 LRIT2 GHITM LOC105378397 RGR CCSER2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |