Assay Details
Target Gene Details
Entrez Gene ID: | 105376488 |
Gene Name: | uncharacterized LOC105376488 |
Gene Aliases: |
- |
Location: |
Chr.10:33334437-33385325 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105376488 | XM_017017010.1 | XP_016872499.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)