Assay Details
Target Gene Details
Entrez Gene ID: | 80313 |
Gene Name: | leucine rich repeat containing 27 |
Gene Aliases: |
- |
Location: |
Chr.10:132330063-132381506 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1054569 | Chr.10:132009350 - 132351508 on Build GRCh38 | Gain | DPYSL4 STK32C JAKMIP3 LRRC27 |
esv3624988 | Chr.10:132338306 - 132366249 on Build GRCh38 | Gain | LRRC27 |
nsv1051555 | Chr.10:132295188 - 132415461 on Build GRCh38 | Gain | PWWP2B STK32C LRRC27 |
esv3624986 | Chr.10:132322882 - 132551350 on Build GRCh38 | Gain | PWWP2B STK32C LOC107984282 INPP5A LOC105378568 C10orf91 LRRC27 |
esv2761642 | Chr.10:132048518 - 132365985 on Build GRCh38 | Gain | DPYSL4 STK32C JAKMIP3 LRRC27 |
nsv1054934 | Chr.10:130334322 - 133158679 on Build GRCh38 | Gain | TCERG1L PWWP2B BNIP3 STK32C LINC01164 ADGRA1-AS1 TCERG1L-AS1 INPP5A ADGRA1 LOC105378568 PPP2R2D JAKMIP3 LINC01166 NKX6-2 MIR378C DPYSL4 LOC107984282 CFAP46 LINC01168 LINC01167 C10orf91 LRRC27 |
nsv1052405 | Chr.10:132336481 - 132390781 on Build GRCh38 | Gain | LRRC27 |