Assay Details
Target Gene Details
Entrez Gene ID: | 79746 |
Gene Name: | enoyl-CoA hydratase domain containing 3 |
Gene Aliases: |
- |
Location: |
Chr.10:11742357-11764070 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ECHDC3 | NM_024693.4 | NP_078969.2 | ||
XM_011519689.1 | XP_011517991.1 | |||
AF275677.1 | AAG24387.1 | |||
AF289604.1 | AAL55788.1 | |||
AK024562.1 | BAB14928.1 | |||
AK222511.1 | BAD96231.1 | |||
AK290902.1 | ||||
BC001091.1 | AAH01091.1 | |||
BC009617.1 | AAH09617.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |