Assay Details
Target Gene Details
Entrez Gene ID: | 1558 |
Gene Name: | cytochrome P450 family 2 subfamily C member 8 |
Gene Aliases: |
CPC8, CYPIIC8, MP-12/MP-20 |
Location: |
Chr.10:95036772-95069512 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CYP2C8 | NM_000770.3 | NP_000761.3 | ||
NM_001198853.1 | NP_001185782.1 | |||
NM_001198854.1 | NP_001185783.1 | |||
NM_001198855.1 | NP_001185784.1 | |||
AK292753.1 | ||||
AK293327.1 | ||||
AK293328.1 | ||||
AK298401.1 | ||||
AK303813.1 | ||||
AK315823.1 | ||||
BC020596.1 | AAH20596.1 | |||
CD014119.1 | ||||
M17397.1 | AAA35739.1 | |||
M17398.1 | AAA35740.1 | |||
M21941.1 | AAA52160.1 | |||
M21942.1 | AAA52161.1 | |||
Y00498.1 | CAA68550.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv551964 | Chr.10:95045614 - 95102529 on Build GRCh38 | Loss | CYP2C8 |
esv3624264 | Chr.10:94923968 - 95099386 on Build GRCh38 | Gain | CYP2C8 CYP2C9 |
nsv1035409 | Chr.10:94735420 - 95072300 on Build GRCh38 | Loss | CYP2C8 CYP2C9 CYP2C18 CYP2C19 |
nsv1044182 | Chr.10:95024239 - 95054932 on Build GRCh38 | Gain | CYP2C8 |
nsv1050544 | Chr.10:95007177 - 95054932 on Build GRCh38 | Gain | CYP2C8 |
esv2761618 | Chr.10:95024251 - 95054932 on Build GRCh38 | Gain | CYP2C8 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |