Assay Details
Target Gene Details
Entrez Gene ID: | 783 |
Gene Name: | calcium voltage-gated channel auxiliary subunit beta 2 |
Gene Aliases: |
CACNLB2, CAVB2, MYSB |
Location: |
Chr.10:18140560-18545881 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CACNB2 | NM_001167945.1 | NP_001161417.1 | ||
NM_201571.3 | NP_963865.2 | |||
NM_201572.3 | NP_963866.2 | |||
NM_201593.2 | NP_963887.2 | |||
NM_201596.2 | NP_963890.2 | |||
NM_201597.2 | NP_963891.1 | |||
AF137376.1 | AAD33729.1 | |||
AF423190.1 | AAL16949.1 | |||
AF423191.1 | AAL16950.1 | |||
AK293837.1 | ||||
AK296471.1 | ||||
AK307526.1 | ||||
AK311694.1 | ||||
AL119895.1 | ||||
AY393858.1 | AAQ97606.1 | |||
AY393860.1 | AAQ97608.1 | |||
AY393861.1 | AAQ97609.1 | |||
AY675091.1 | AAV49501.1 | |||
BC136409.1 | ||||
DC307859.1 | ||||
DC352526.1 | ||||
S60415.1 | AAB51370.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |