Assay Details
Target Gene Details
Entrez Gene ID: | 83592 |
Gene Name: | aldo-keto reductase family 1 member E2 |
Gene Aliases: |
AKR1CL2, AKRDC1, LoopADR, hTSP, htAKR |
Location: |
Chr.10:4824554-4908421 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1045155 | Chr.10:4830047 - 5169355 on Build GRCh38 | Loss | AKR1C1 AKR1E2 AKR1C8P AKR1C2 AKR1C3 AKR1C6P |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |