Assay Details
Target Gene Details
Entrez Gene ID: | 10210 |
Gene Name: | TOP1 binding arginine/serine rich protein |
Gene Aliases: |
LUN, P53BP3, RP31, TP53BPL |
Location: |
Chr.9:32540544-32552628 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TOPORS | NM_001195622.1 | NP_001182551.1 | ||
NM_005802.4 | NP_005793.2 | |||
AB045732.1 | BAB03714.1 | |||
AB045733.1 | BAB03715.1 | |||
AF098300.1 | AAD23379.1 | |||
AU280552.1 | ||||
BC060884.1 | AAH60884.1 | |||
DA238762.1 | ||||
U82939.1 | AAC98530.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831540 | Chr.9:32509387 - 32651746 on Build GRCh38 | Gain | TOPORS TOPORS-AS1 TAF1L DDX58 NDUFB6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |