Assay Details
Target Gene Details
Entrez Gene ID: | 22949 |
Gene Name: | prostaglandin reductase 1 |
Gene Aliases: |
LTB4DH, PGR1, ZADH3 |
Location: |
Chr.9:111549722-111599855 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTGR1 | NM_001146109.1 | NP_001139581.1 | ||
AK298379.1 |
Target Gene Details
Entrez Gene ID: | 158399 |
Gene Name: | zinc finger protein 483 |
Gene Aliases: |
ZKSCAN16, ZSCAN48 |
Location: |
Chr.9:111525159-111577844 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZNF483 | NM_001007169.3 | NP_001007170.1 | ||
AK092811.1 | BAC03981.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |