Assay Details
Target Gene Details
Entrez Gene ID: | 256691 |
Gene Name: | MAM domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.9:70043519-70226972 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MAMDC2 | NM_153267.4 | NP_694999.3 | ||
NR_125850.1 | ||||
XM_011518475.2 | XP_011516777.1 | |||
AK304040.1 | ||||
AK304531.1 | ||||
BC015417.2 | AAH15417.1 | |||
BC016383.1 | AAH16383.1 | |||
BC063634.1 | AAH63634.1 | |||
BX538309.1 | 1 | 1679 | CAD98089.1 |
Target Gene Details
Entrez Gene ID: | 100507299 |
Gene Name: | SMC5 antisense RNA 1 (head to head) |
Gene Aliases: |
MAMDC2-AS2 |
Location: |
Chr.9:70216059-70258874 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SMC5-AS1 | NR_039990.1 | |||
AK124136.1 | ||||
CR936772.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1047862 | Chr.9:70131959 - 70523371 on Build GRCh38 | Gain | SMC5 SMC5-AS1 MAMDC2-AS1 KLF9 MAMDC2 |