Assay Details
Target Gene Details
Entrez Gene ID: | 1621 |
Gene Name: | dopamine beta-hydroxylase |
Gene Aliases: |
DBM |
Location: |
Chr.9:133636363-133659344 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DBH | NM_000787.3 | NP_000778.3 | ||
BC017174.2 | AAH17174.1 | |||
CD014125.1 | ||||
X13255.1 | CAA31631.1 | |||
X13256.1 | CAA31632.1 | |||
Y00096.1 | CAA68285.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv615639 | Chr.9:133639476 - 133654580 on Build GRCh38 | Loss | DBH |
esv2739138 | Chr.9:132727483 - 135521553 on Build GRCh38 | Deletion | STKLD1 LINC00094 SNORD24 SARDH LOC100996574 MIR3689E RXRA FCN2 MIR3689C PPP1R26-AS1 C9orf62 CACFD1 SURF2 SPACA9 MRPS2 ABO LOC100130548 LOC105376306 LOC101928193 MIR548AW PPP1R26 BRD3 GFI1B RALGDS CEL VAV2 DBH-AS1 RNU6ATAC C9orf116 CELP MED22 MIR3689F SNORD36C DBH MIR4669 MIR3689A MIR3689D1 REXO4 LOC101448202 TSC1 OLFM1 AK8 LCN1 MIR6877 ADAMTSL2 SNORD141A RPL7A SURF6 MIR3689D2 LOC401557 SLC2A6 COL5A1-AS1 FCN1 COL5A1 MIR3689B ADAMTS13 SNORD36B OBP2B SURF4 GBGT1 SNORD36A SURF1 LOC101928525 TMEM8C FAM163B WDR5 GTF3C5 |
nsv615638 | Chr.9:133633021 - 133640119 on Build GRCh38 | Loss | DBH |
nsv951800 | Chr.9:133627979 - 133696678 on Build GRCh38 | Deletion | DBH-AS1 SARDH DBH |
nsv469906 | Chr.9:133629444 - 133643961 on Build GRCh38 | Loss | DBH |
esv3576711 | Chr.9:133582804 - 133778331 on Build GRCh38 | Gain | DBH-AS1 SARDH DBH VAV2 FAM163B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |