Assay Details
Target Gene Details
Entrez Gene ID: | 23245 |
Gene Name: | astrotactin 2 |
Gene Aliases: |
bA67K19.1 |
Location: |
Chr.9:116425225-117415039 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ASTN2 | NM_001184734.1 | NP_001171663.1 | ||
NM_014010.4 | NP_054729.3 | |||
NM_198186.3 | NP_937829.3 | |||
NM_198187.3 | NP_937830.3 | |||
NM_198188.2 | NP_937831.1 | |||
AB014534.1 | ||||
AF116574.1 | AAF14357.1 | |||
AK024064.1 | ||||
AK127350.1 | ||||
AK315025.1 | ||||
BC018759.2 | AAH18759.2 | |||
BC029272.1 | AAH29272.1 | |||
BC093835.1 | ||||
BC101667.1 | ||||
BC143289.1 | ||||
BC143292.1 | ||||
BC143293.1 | ||||
BC146756.1 | ||||
BM552790.1 | ||||
BX649176.1 |
Target Gene Details
Entrez Gene ID: | 100128505 |
Gene Name: | ASTN2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.9:116504283-116562293 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ASTN2-AS1 | NR_033973.1 | |||
AK057447.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831701 | Chr.9:116485758 - 116642973 on Build GRCh38 | Gain | ASTN2-AS1 ASTN2 |
nsv615254 | Chr.9:116500349 - 116577184 on Build GRCh38 | Loss | ASTN2-AS1 ASTN2 |
esv3891719 | Chr.9:116525276 - 116552571 on Build GRCh38 | Loss | ASTN2-AS1 ASTN2 |
esv2761554 | Chr.9:116541782 - 116595630 on Build GRCh38 | Loss | ASTN2-AS1 ASTN2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |