Assay Details
Target Gene Details
Entrez Gene ID: | 1666 |
Gene Name: | 2,4-dienoyl-CoA reductase 1, mitochondrial |
Gene Aliases: |
DECR, NADPH, SDR18C1 |
Location: |
Chr.8:90001337-90052004 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DECR1 | NM_001359.1 | NP_001350.1 | ||
XM_005250808.4 | XP_005250865.1 | |||
XM_011516880.2 | XP_011515182.1 | |||
XM_017013147.1 | XP_016868636.1 | |||
XM_017013148.1 | XP_016868637.1 | |||
AK300069.1 | ||||
BC105080.1 | AAI05081.1 | |||
BC105082.1 | AAI05083.1 | |||
L26050.1 | AAA67551.1 | |||
U49352.1 | AAB09423.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759628 | Chr.8:89988558 - 90317441 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)