Assay Details
Target Gene Details
Entrez Gene ID: | 7534 |
Gene Name: | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein zeta |
Gene Aliases: |
14-3-3-zeta, HEL-S-3, HEL-S-93, HEL4, KCIP-1, YWHAD |
Location: |
Chr.8:100918576-100954068 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831409 | Chr.8:100924261 - 101081205 on Build GRCh38 | Loss | LOC401471 YWHAZ FLJ42969 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |