Assay Details
Target Gene Details
Entrez Gene ID: | 79660 |
Gene Name: | protein phosphatase 1 regulatory subunit 3B |
Gene Aliases: |
GL, PPP1R4, PTG |
Location: |
Chr.8:9136254-9151642 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPP1R3B | NM_001201329.1 | NP_001188258.1 | ||
NM_024607.3 | NP_078883.2 | |||
XM_006716253.3 | XP_006716316.1 | |||
XM_017013835.1 | XP_016869324.1 | |||
AK024067.1 | BAB14811.1 | |||
AK096119.1 | ||||
BC043388.1 | AAH43388.1 | |||
BQ431880.1 | ||||
DA100998.1 | ||||
DB282578.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv610206 | Chr.8:9103507 - 9304538 on Build GRCh38 | Loss | LOC101929128 PPP1R3B |
esv3616157 | Chr.8:8901663 - 9165103 on Build GRCh38 | Loss | PPP1R3B ERI1 MIR4660 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |