Assay Details
Target Gene Details
Entrez Gene ID: | 22882 |
Gene Name: | zinc fingers and homeoboxes 2 |
Gene Aliases: |
AFR1, RAF |
Location: |
Chr.8:122781349-122974515 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZHX2 | NM_014943.3 | NP_055758.1 | ||
XM_005250837.4 | XP_005250894.1 | |||
AB020661.1 | ||||
BC042145.1 | AAH42145.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1032827 | Chr.8:122655842 - 123258160 on Build GRCh38 | Gain | FAM83A-AS1 ZHX1-C8orf76 LINC01151 MIR4663 C8orf76 DERL1 ZHX2 FAM83A ZHX1 TBC1D31 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |