Assay Details
Target Gene Details
Entrez Gene ID: | 10499 |
Gene Name: | nuclear receptor coactivator 2 |
Gene Aliases: |
GRIP1, KAT13C, NCoA-2, SRC2, TIF2, bHLHe75 |
Location: |
Chr.8:70109762-70405390 on Build GRCh38 |
Assay Gene Location: | Within Intron 30 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831352 | Chr.8:69999548 - 70182606 on Build GRCh38 | Gain | NCOA2 PRDM14 |
nsv6247 | Chr.8:70118114 - 70163017 on Build GRCh38 | Deletion | NCOA2 |
nsv6246 | Chr.8:70106516 - 70137048 on Build GRCh38 | Insertion | NCOA2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |