Assay Details
Target Gene Details
Entrez Gene ID: | 9258 |
Gene Name: | malignant fibrous histiocytoma amplified sequence 1 |
Gene Aliases: |
LRRC65, MASL1 |
Location: |
Chr.8:8783354-8893621 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MFHAS1 | NM_004225.2 | NP_004216.2 | ||
AB016816.1 | BAA74737.1 | |||
AK027007.1 | ||||
BC014226.2 | AAH14226.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv8249 | Chr.8:8817893 - 8826139 on Build GRCh38 | Loss | MFHAS1 |
nsv831228 | Chr.8:8750877 - 8933280 on Build GRCh38 | Loss | MFHAS1 ERI1 |
nsv1018828 | Chr.8:8752370 - 8933150 on Build GRCh38 | Loss | MFHAS1 ERI1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |