Assay Details
Target Gene Details
Entrez Gene ID: | 81034 |
Gene Name: | solute carrier family 25 member 32 |
Gene Aliases: |
MFT, MFTC, RREI |
Location: |
Chr.8:103398638-103415335 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC25A32 | NM_030780.4 | NP_110407.2 | ||
NR_102337.1 | ||||
NR_102338.1 | ||||
XM_017013877.1 | XP_016869366.1 | |||
AF283645.1 | AAG37834.1 | |||
AK027531.1 | BAB55180.1 | |||
AK027787.1 | BAB55368.1 | |||
AK125492.1 | ||||
AK300748.1 | ||||
AL046268.1 | ||||
BC021893.1 | AAH21893.1 | |||
BP274577.1 | ||||
DB087013.1 | ||||
DC331449.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv611832 | Chr.8:103388580 - 103469507 on Build GRCh38 | Loss | SLC25A32 DCAF13 |
dgv7279n100 | Chr.8:103349626 - 103867456 on Build GRCh38 | Gain | SLC25A32 CTHRC1 DCAF13 RIMS2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |