Assay Details
Target Gene Details
Entrez Gene ID: | 102725541 |
Gene Name: | EGFR long non-coding downstream RNA |
Gene Aliases: |
Fabl, LINC01156 |
Location: |
Chr.7:55238436-55255635 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ELDR | NR_110426.1 | |||
DB084610.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2734425 | Chr.7:53868335 - 55917254 on Build GRCh38 | Deletion | LOC101060341 ELDR LOC100996654 VSTM2A HPVC1 SEC61G VSTM2A-OT1 LOC107986797 LINC01445 EGFR-AS1 PSPHP1 ZNF713 LANCL2 SEPT14 VOPP1 EGFR FKBP9P1 |