Assay Details
Target Gene Details
Entrez Gene ID: | 4850 |
Gene Name: | CCR4-NOT transcription complex subunit 4 |
Gene Aliases: |
CLONE243, NOT4, NOT4H |
Location: |
Chr.7:135361795-135510127 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CNOT4 | NM_001190847.1 | NP_001177776.1 | ||
NM_001190849.1 | NP_001177778.1 | |||
NM_001190850.1 | NP_001177779.1 | |||
NM_013316.3 | NP_037448.2 | |||
XM_017012235.1 | XP_016867724.1 | |||
AF091094.1 | AAC72963.1 | |||
AK074493.1 | ||||
AK074671.1 | BAC11125.1 | |||
AK300365.1 | ||||
AK314540.1 | ||||
CA396599.1 | ||||
U71267.1 | AAD00179.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv29990 | Chr.7:135328379 - 135372394 on Build GRCh38 | Gain | CNOT4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |