Assay Details
Target Gene Details
Entrez Gene ID: | 83698 |
Gene Name: | calneuron 1 |
Gene Aliases: |
CABP8 |
Location: |
Chr.7:71779491-72504271 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv516049 | Chr.7:71561478 - 71815314 on Build GRCh38 | Gain | CALN1 WBSCR17 |
nsv607459 | Chr.7:71774529 - 71801548 on Build GRCh38 | Gain | CALN1 |
esv3613640 | Chr.7:71778998 - 71802508 on Build GRCh38 | Gain | CALN1 |
nsv1033922 | Chr.7:71750208 - 71802242 on Build GRCh38 | Gain | CALN1 |
nsv1017575 | Chr.7:70687626 - 71905471 on Build GRCh38 | Gain | MIR3914-2 AUTS2 CALN1 WBSCR17 MIR3914-1 |
dgv6456n100 | Chr.7:71568840 - 71823976 on Build GRCh38 | Gain | CALN1 WBSCR17 |
nsv1026896 | Chr.7:71538477 - 72256531 on Build GRCh38 | Gain | CALN1 WBSCR17 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |