Assay Details
Target Gene Details
Entrez Gene ID: | 10643 |
Gene Name: | insulin like growth factor 2 mRNA binding protein 3 |
Gene Aliases: |
CT98, IMP-3, IMP3, KOC, KOC1, VICKZ3 |
Location: |
Chr.7:23310209-23470674 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IGF2BP3 | NM_006547.2 | NP_006538.2 | ||
XM_006715639.2 | XP_006715702.1 | |||
XM_011515089.2 | XP_011513391.1 | |||
XM_011515090.2 | XP_011513392.1 | |||
XM_011515091.2 | XP_011513393.1 | |||
XM_011515092.2 | XP_011513394.1 | |||
XM_011515093.2 | XP_011513395.1 | |||
AF117108.1 | AAD09828.1 | |||
BC065269.1 | AAH65269.1 | |||
BX640800.1 | CAE45883.1 | |||
BX648488.1 | CAH56186.1 | |||
U76705.1 | AAD09223.1 | |||
U97188.1 | AAC35208.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv606410 | Chr.7:23293751 - 23343516 on Build GRCh38 | Loss |
![]() ![]() |
nsv1016316 | Chr.7:23221900 - 23318018 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)