Assay Details
Target Gene Details
Entrez Gene ID: | 22853 |
Gene Name: | lemur tyrosine kinase 2 |
Gene Aliases: |
AATYK2, BREK, KPI-2, KPI2, LMR2, PPP1R100, cprk, hBREK |
Location: |
Chr.7:98106868-98209633 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LMTK2 | NM_014916.3 | NP_055731.2 | ||
XM_011515981.2 | XP_011514283.1 | |||
AB029002.1 | BAA83031.2 | |||
AY130988.1 | AAN08717.1 | |||
BC131504.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831066 | Chr.7:98097567 - 98261238 on Build GRCh38 | Loss | TECPR1 LMTK2 BHLHA15 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |