Assay Details
Target Gene Details
Entrez Gene ID: | 5444 |
Gene Name: | paraoxonase 1 |
Gene Aliases: |
ESA, MVCD5, PON |
Location: |
Chr.7:95298357-95324572 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PON1 | NM_000446.5 | NP_000437.3 | ||
AK314027.1 | ||||
BC074719.2 | AAH74719.1 | |||
D84371.1 | BAA12327.1 | |||
M63012.1 | AAB59538.1 | |||
M63013.1 | AAA60142.1 | |||
M63014.1 | AAA60143.1 | |||
S64615.1 | AAB27714.2 | |||
U53784.1 | AAA97957.1 | |||
Z70723.1 | CAA94728.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1023193 | Chr.7:93602376 - 95828660 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)