Assay Details
Target Gene Details
Entrez Gene ID: | 23382 |
Gene Name: | adenosylhomocysteinase like 2 |
Gene Aliases: |
ADOHCYASE3, IRBIT2 |
Location: |
Chr.7:129225014-129430211 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AHCYL2 | NM_001130720.2 | NP_001124192.1 | ||
NM_015328.3 | NP_056143.1 | |||
XM_017011906.1 | XP_016867395.1 | |||
AB020635.1 | ||||
BC008349.1 | AAH08349.1 | |||
BC024325.1 | AAH24325.1 | |||
BG700017.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831127 | Chr.7:129076202 - 129234850 on Build GRCh38 | Gain | SMO TSPAN33 AHCYL2 LOC407835 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |