Assay Details
Target Gene Details
Entrez Gene ID: | 346653 |
Gene Name: | family with sequence similarity 71 member F2 |
Gene Aliases: |
FAM137B |
Location: |
Chr.7:128671628-128702262 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM71F2 | XM_011516134.2 | XP_011514436.1 | ||
XM_011516135.2 | XP_011514437.1 | |||
XM_011516136.2 | XP_011514438.1 | |||
XM_017012147.1 | XP_016867636.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2670312 | Chr.7:128499519 - 128999613 on Build GRCh38 | Deletion | IRF5 FAM71F1 METTL2B LINC01000 LOC100130705 CCDC136 OPN1SW TNPO3 CALU FLNC KCP FAM71F2 ATP6V1F |
esv2735137 | Chr.7:128499440 - 128999664 on Build GRCh38 | Deletion | IRF5 FAM71F1 METTL2B LINC01000 LOC100130705 CCDC136 OPN1SW TNPO3 CALU FLNC KCP FAM71F2 ATP6V1F |