Assay Details
Target Gene Details
Entrez Gene ID: | 84792 |
Gene Name: | family with sequence similarity 220 member A |
Gene Aliases: |
ACPIN1, C7orf70, SIPAR |
Location: |
Chr.7:6329409-6348959 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM220A | NM_001037163.1 | NP_001032240.1 | ||
AF533707.1 | AAQ09020.1 | |||
AK093156.1 | BAC04076.1 | |||
AK311878.1 | ||||
BC006110.1 | ||||
BC140712.1 | ||||
BC142977.1 | ||||
BP354450.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1020236 | Chr.7:6209299 - 6650746 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1025381 | Chr.7:6317073 - 6370187 on Build GRCh38 | Loss |
![]() |
nsv436904 | Chr.7:5035574 - 6713282 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)