Assay Details
Target Gene Details
Entrez Gene ID: | 94239 |
Gene Name: | H2A histone family member V |
Gene Aliases: |
H2A.Z-2, H2AV |
Location: |
Chr.7:44826889-44848126 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
H2AFV | NM_012412.4 | NP_036544.1 | ||
NM_138635.3 | NP_619541.1 | |||
NM_201436.2 | NP_958844.1 | |||
NM_201516.2 | NP_958924.1 | |||
NM_201517.2 | NP_958925.1 | |||
AB209001.1 | BAD92238.1 | |||
AF081192.1 | AAC31938.1 | |||
AK023973.1 | ||||
AK129989.1 | ||||
AL520811.3 | ||||
BC000098.2 | AAH00098.1 | |||
BC004274.1 | ||||
BC014885.2 | AAH14885.1 | |||
BC070169.1 | AAH70169.1 | |||
BF242439.1 | ||||
BG754026.1 | ||||
BG773256.1 | ||||
BI257891.1 | ||||
BM452844.1 | ||||
BM453663.1 | ||||
CB148943.1 | ||||
DB460745.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830985 | Chr.7:44793475 - 44979588 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)