Assay Details
Target Gene Details
Entrez Gene ID: | 101927870 |
Gene Name: | uncharacterized LOC101927870 |
Gene Aliases: |
- |
Location: |
Chr.7:103445207-103514007 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101927870 | NR_110141.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3576411 | Chr.7:103468275 - 103474748 on Build GRCh38 | Gain | RELN LOC101927870 |