Assay Details
Target Gene Details
Entrez Gene ID: | 101927870 |
Gene Name: | uncharacterized LOC101927870 |
Gene Aliases: |
- |
Location: |
Chr.7:103445207-103514007 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101927870 | NR_110141.1 |
Target Gene Details
Entrez Gene ID: | 5649 |
Gene Name: | reelin |
Gene Aliases: |
ETL7, LIS2, PRO1598, RL |
Location: |
Chr.7:103471784-103989516 on Build GRCh38 |
Assay Gene Location: | Within Intron 63 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RELN | NM_005045.3 | NP_005036.2 | ||
NM_173054.2 | NP_774959.1 | |||
AK293863.1 | ||||
AK297459.1 | ||||
U79716.1 | AAC51105.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |