Assay Details
Target Gene Details
Entrez Gene ID: | 55536 |
Gene Name: | cell division cycle associated 7 like |
Gene Aliases: |
JPO2, R1, RAM2 |
Location: |
Chr.7:21900899-21945924 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CDCA7L | NM_001127370.2 | NP_001120842.1 | ||
NM_001127371.2 | NP_001120843.1 | |||
NM_018719.4 | NP_061189.2 | |||
AK022955.1 | BAB14330.1 | |||
AK095965.1 | ||||
AK292490.1 | ||||
AK300781.1 | ||||
AY161168.1 | AAO17570.1 | |||
AY161169.1 | AAO17571.1 | |||
BC009352.2 | AAH09352.2 | |||
BC014630.2 | AAH14630.2 | |||
BC025242.1 | AAH25242.1 | |||
BC032576.1 | AAH32576.1 | |||
BC039823.1 | AAH39823.1 | |||
BX422040.2 | ||||
DA041070.1 | ||||
DC331792.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830922 | Chr.7:21753768 - 21925057 on Build GRCh38 | Loss |
![]() ![]() |
nsv1027453 | Chr.7:21878856 - 21942830 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)