Assay Details
Target Gene Details
Entrez Gene ID: | 6604 |
Gene Name: | SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 3 |
Gene Aliases: |
BAF60C, CRACD3, Rsc6p |
Location: |
Chr.7:151238973-151277436 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SMARCD3 | NM_001003802.1 | NP_001003802.1 | ||
NM_003078.3 | NP_003069.2 | |||
XM_011516521.2 | XP_011514823.1 | |||
XM_017012556.1 | XP_016868045.1 | |||
AK127595.1 | ||||
AY450431.1 | AAR88511.1 | |||
BC002628.2 | AAH02628.1 | |||
BC110350.1 | AAI10351.1 | |||
BI598614.1 | ||||
BX648385.1 | ||||
U66619.1 | AAC50697.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv609035 | Chr.7:151199137 - 151258679 on Build GRCh38 | Loss | ABCF2 CHPF2 IQCA1L MIR671 SMARCD3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |