Assay Details
Target Gene Details
Entrez Gene ID: | 219285 |
Gene Name: | sterile alpha motif domain containing 9 like |
Gene Aliases: |
C7orf6, DRIF2, UEF1 |
Location: |
Chr.7:93130054-93148401 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SAMD9L | NM_001303496.1 | NP_001290425.1 | ||
NM_001303497.1 | NP_001290426.1 | |||
NM_001303498.1 | NP_001290427.1 | |||
NM_001303500.1 | NP_001290429.1 | |||
NM_152703.3 | NP_689916.2 | |||
XM_005250193.3 | XP_005250250.1 | |||
XM_006715890.1 | XP_006715953.1 | |||
XM_011515903.2 | XP_011514205.1 | |||
XM_017011822.1 | XP_016867311.1 | |||
XM_017011823.1 | XP_016867312.1 | |||
AB095926.1 | ||||
AF474973.1 | AAQ05771.1 | |||
AY195582.1 | AAP57709.1 | |||
AY195583.1 | AAP57710.1 | |||
AY195584.1 | AAP57711.1 | |||
AY195585.1 | AAP57712.1 | |||
AY195586.1 | AAP57713.1 | |||
AY195587.1 | ||||
BC127117.1 | ||||
BC127118.1 | ||||
DQ068177.1 |