Assay Details
Target Gene Details
Entrez Gene ID: | 23553 |
Gene Name: | hyaluronoglucosaminidase 4 |
Gene Aliases: |
CSHY |
Location: |
Chr.7:123763708-123877481 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HYAL4 | XM_017011911.1 | XP_016867400.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3891198 | Chr.7:121885430 - 123886905 on Build GRCh38 | Gain | PTPRZ1 LMOD2 CADPS2 LOC102724555 ASB15 HYAL4 IQUB FEZF1-AS1 WASL TAS2R16 HYALP1 RNF148 SLC13A1 AASS RNF133 NDUFA5 FEZF1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |